Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025407.2 | NP_079683.2 | 11 - 12 | 1371 | 96 | |
GenBank mRNA | AI462428.1 | - | 2 - 3 | 96 | ||
AK002292.1 | - | 11 - 12 | 1308 | 96 | ||
AK004031.1 | - | 11 - 12 | 1317 | 96 | ||
AK010553.1 | - | 11 - 12 | 1311 | 96 | ||
AK013128.1 | - | 11 - 12 | 1311 | 96 | ||
AK151764.1 | - | 11 - 12 | 1313 | 96 | ||
AK160337.1 | - | 11 - 12 | 1318 | 96 | ||
AK167911.1 | - | 11 - 12 | 1311 | 96 | ||
AK168218.1 | - | 11 - 12 | 1313 | 96 | ||
BC018405.1 | - | 4 - 5 | 424 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |