Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trnau1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027925.3 | NP_082201.2 | 6 - 7 | 691 | 95 | |
GenBank mRNA | AK003534.1 | - | 5 - 6 | 1354 | 95 | |
AK152986.1 | - | 6 - 7 | 640 | 95 | ||
BC005795.1 | - | 6 - 7 | 648 | 95 | ||
BC048840.1 | - | 6 - 7 | 642 | 5008141 | 95 | |
BC055454.1 | - | 6 - 7 | 654 | 4014366 | 95 | |
BC099594.1 | - | 4 - 5 | 376 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |