Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028607.1 | NP_082883.1 | 3 - 4 | 329 | 85 | |
GenBank mRNA | AK010745.1 | - | 3 - 4 | 326 | 85 | |
AK145313.1 | - | 3 - 4 | 330 | 85 | ||
BC065806.1 | - | 3 - 4 | 322 | 1547643 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |