Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133854.3 | NP_598615.1 | 2 - 3 | 235 | 86 | |
GenBank mRNA | AF086838.1 | - | 2 - 3 | 196 | 86 | |
AK007458.1 | - | 2 - 3 | 216 | 86 | ||
AK035147.1 | - | 2 - 3 | 232 | 86 | ||
AK048007.1 | - | 2 - 3 | 224 | 86 | ||
AK050151.1 | - | 2 - 3 | 236 | 86 | ||
AK077840.1 | - | 2 - 3 | 226 | 86 | ||
AK080718.1 | - | 2 - 3 | 226 | 86 | ||
AK086994.1 | - | 1 - 2 | 167 | 86 | ||
AK135006.1 | - | 2 - 3 | 227 | 86 | ||
AK152057.1 | - | 2 - 3 | 204 | 86 | ||
BC048691.1 | - | 2 - 3 | 207 | 6511395 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |