Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252568.2 | NP_001239497.1 | 2 - 3 | 509 | 75 | |
NM_001252570.1 | NP_001239499.1 | 1 - 2 | 130 | 75 | ||
NM_001252571.1 | NP_001239500.1 | 1 - 2 | 130 | 75 | ||
NM_001281829.1 | NP_001268758.1 | 2 - 3 | 156 | 75 | ||
NM_172267.4 | NP_758471.1 | 2 - 3 | 509 | 75 | ||
GenBank mRNA | AI182929.1 | - | 2 - 3 | 131 | 75 | |
AK005293.1 | - | 2 - 3 | 510 | 75 | ||
AK046118.1 | - | 1 - 2 | 99 | 75 | ||
AK138607.1 | - | 2 - 3 | 184 | 75 | ||
BC039982.1 | - | 2 - 3 | 151 | 75 | ||
BC132271.1 | - | 2 - 3 | 120 | 40130548 | 75 | |
BC132273.1 | - | 2 - 3 | 120 | 40130550 | 75 | |
BC145356.1 | - | 2 - 3 | 120 | 9053914 | 75 | |
BC171925.1 | - | 2 - 3 | 120 | 75 | ||
BQ829083.1 | - | 1 - 2 | 126 | 75 | ||
BY196857.1 | - | 2 - 3 | 158 | 75 | ||
BY344206.1 | - | 2 - 3 | 163 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |