Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172277.2 | NP_758481.1 | 5 - 6 | 611 | 97 | |
GenBank mRNA | AK045061.1 | - | 5 - 6 | 604 | 97 | |
AK082871.1 | - | 5 - 6 | 604 | 97 | ||
AK148429.1 | - | 5 - 6 | 604 | 97 | ||
AK156817.1 | - | 5 - 6 | 557 | 97 | ||
AK161672.1 | - | 4 - 5 | 545 | 97 | ||
BC019142.1 | - | 4 - 5 | 607 | 97 | ||
BC037599.1 | - | 5 - 6 | 637 | 5150754 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |