Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172277.2 | NP_758481.1 | 9 - 10 | 1122 | 104 | |
GenBank mRNA | AK045061.1 | - | 9 - 10 | 1115 | 104 | |
AK082871.1 | - | 9 - 10 | 1115 | 104 | ||
AK148429.1 | - | 9 - 10 | 1115 | 104 | ||
AK156817.1 | - | 9 - 10 | 1068 | 104 | ||
BC019142.1 | - | 8 - 9 | 1118 | 104 | ||
BC037599.1 | - | 9 - 10 | 1148 | 5150754 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |