Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177850.3 | NP_808518.1 | 7 - 8 | 805 | 111 | |
GenBank mRNA | AK033770.1 | - | 7 - 8 | 807 | 111 | |
AY363993.1 | - | 7 - 8 | 803 | 111 | ||
AY648037.1 | - | 7 - 8 | 805 | 111 | ||
BC118049.1 | - | 7 - 8 | 799 | 40106697 | 111 | |
BC118504.1 | - | 7 - 8 | 799 | 40106694 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |