Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10403942 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10408240 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10494403 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10408248 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10494400 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10404050 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10404029 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10494406 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10404066 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10408084 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10408203 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10494405 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10494402 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10404065 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10494395 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0300007471.mm.1 |
TC0300000934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007468.mm.1 |
TC0300000934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007469.mm.1 |
NM_178215.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007469.mm.1 |
TC0300000934.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1453573_at |
- |
- |
Mouse Expression Set 430 |
1460314_s_at |
- |
- |
Mouse Expression Set 430 |
1418367_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1418366_at |
- |
- |
Mouse Expression Set 430 |
1418367_x_at |
- |
- |
Mouse Expression Set 430 |
1460314_PM_s_at |
- |
- |
Mouse Expression Set 430 |
1427864_PM_at |
- |
- |
Mouse Expression Set 430 |
1418366_PM_at |
- |
- |
Mouse Expression Set 430 |
1453573_PM_at |
- |
- |
Mouse Expression Set 430 |
1427864_at |
- |
- |
Mouse Expression Set 430 |
1442051_at |
- |
- |
Mouse Expression Set 430 |
1442051_PM_at |
- |
- |
Murine Genome U74v2 Set |
94756_at |
- |
- |
Murine Genome U74v2 Set |
93023_f_at |
- |
- |
Murine Genome U74v2 Set |
96416_f_at |
- |
- |
Murine Genome U74v2 Set |
93068_r_at |
- |
- |
Murine Genome U74v2 Set |
93067_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
96416_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
93022_PM_i_at |
- |
- |
Murine Genome U74v2 Set |
93067_f_at |
- |
- |
Murine Genome U74v2 Set |
94756_PM_at |
- |
- |
Murine Genome U74v2 Set |
93022_i_at |
- |
- |
Murine Genome U74v2 Set |
93068_PM_r_at |
- |
- |
Murine Genome U74v2 Set |
93023_PM_f_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1427864_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1418367_PM_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1453573_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1427864_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1460314_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1418366_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1418367_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1418366_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1453573_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1460314_PM_s_at |
- |
- |