Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akirin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023423.3 | NP_075912.2 | 1 | 55 | 96 | |
GenBank mRNA | AK159632.1 | - | 1 | 43 | 96 | |
AK168530.1 | - | 1 | 37 | 96 | ||
BC003291.1 | - | 1 | 101 | 3498569 | 96 | |
BY315833.1 | - | 1 | 58 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |