Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400004494.mm.1 |
NM_013632.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004494.mm.1 |
NM_013632.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004494.mm.1 |
TC1400000574.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004495.mm.1 |
NM_013632.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004495.mm.1 |
NM_013632.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004495.mm.1 |
TC1400000574.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004491.mm.1 |
NM_013632.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004491.mm.1 |
NM_013632.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004491.mm.1 |
TC1400000574.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004492.mm.1 |
NM_013632.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004492.mm.1 |
NM_013632.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004492.mm.1 |
TC1400000574.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400004493.mm.1 |
NM_013632.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004493.mm.1 |
NM_013632.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400004493.mm.1 |
TC1400000574.mm.1 |
MTA TC |