Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200019412.mm.1 |
NM_173396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019412.mm.1 |
TC0200002401.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019416.mm.1 |
NM_173396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019416.mm.1 |
TC0200002401.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019411.mm.1 |
NM_173396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019411.mm.1 |
TC0200002401.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019413.mm.1 |
NM_173396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019413.mm.1 |
TC0200002401.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019414.mm.1 |
NM_173396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019414.mm.1 |
TC0200002401.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019402.mm.1 |
NM_173396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019402.mm.1 |
TC0200002401.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019415.mm.1 |
NM_173396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019415.mm.1 |
TC0200002401.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019406.mm.1 |
NM_173396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019406.mm.1 |
TC0200002401.mm.1 |
MTA TC |