Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snord17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030762.1 | 1 | 155 | 74 | ||
GenBank mRNA | AF357317.1 | - | 1 | 155 | 74 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
barrier to autointegration factor 2, opposite strand |
Banf2os |
Gm12357, OTTMUSG00000006417 |
-58.1kb | Sense | Chr. 2 - 144059496 - 144207860 on NCBI Build GRCm38 |
Downstream | mitochondrial genome maintainance exonuclease 1 | Mgme1 |
8430406I07Rik, AI426476 |
4.7kb | Anti-sense | Chr. 2 - 144270856 - 144281232 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |