Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spint3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177401.1 | NP_001170872.1 | 1 - 2 | 236 | 99 | |
GenBank mRNA | AK136730.1 | - | 1 - 2 | 214 | 99 | |
BU938823.1 | - | 1 - 2 | 239 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus Probe spans exons |