Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apela | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297554.1 | NP_001284483.1 | 1 - 2 | 57 | 84 | |
GenBank mRNA | AK014119.1 | - | 1 - 2 | 59 | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tripartite motif-containing 61 | Trim61 |
2czf61, AU018704, C85201, E330039K03Rik, Rnf35 |
-18.2kb | Sense | Chr. 8 - 65012976 - 65018688 on NCBI Build GRCm38 |
Downstream | cDNA sequence BC030870 | BC030870 | 48.3kb | Sense | Chr. 8 - 65085613 - 65129537 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |