Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045485.1 | 1 - 2 | 212 | 65 | ||
GenBank mRNA | AK019246.1 | - | 1 - 2 | 148 | 65 | |
AK189495.1 | - | 1 - 2 | 60 | 65 | ||
CK031473.1 | - | 1 - 2 | 159 | 65 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SH2 domain containing 3C | Sh2d3c |
Chat, Nsp3, Shep1 |
-2.2kb | Sense | Chr. 2 - 32721055 - 32755007 on NCBI Build GRCm38 |
Downstream | peptidyl-tRNA hydrolase 1 homolog (S. cerevisiae) | Ptrh1 |
2210013M04Rik, AW046744 |
16.8kb | Sense | Chr. 2 - 32775738 - 32777593 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |