Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045575.1 | 4 - 5 | 348 | 71 | ||
GenBank mRNA | AK134822.1 | - | 4 - 5 | 305 | 71 | |
CK625955.1 | - | 3 - 4 | 291 | 71 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | armadillo repeat containing 1 | Armc1 |
2310016N05Rik, 2900046P06Rik, 3110009G21Rik, AA409258, AW048872, Arcp, C330014L16Rik |
-45.4kb | Anti-sense | Chr. 3 - 19132144 - 19163090 on NCBI Build GRCm38 |
Downstream | phosphodiesterase 7A | Pde7a |
AU015378, AW047537 |
11.5kb | Anti-sense | Chr. 3 - 19223108 - 19311374 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |