Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vezf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016686.4 | NP_057895.2 | 4 - 5 | 1103 | 148 | |
GenBank mRNA | AF104410.3 | - | 4 - 5 | 1103 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |