Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195033.1 | NP_001181962.1 | 6 - 7 | 524 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |