Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00007549.mm.1 |
NR_073451.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007549.mm.1 |
NR_073452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007549.mm.1 |
TC0X00001178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007526.mm.1 |
NM_001271809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007526.mm.1 |
NM_001271810.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007526.mm.1 |
NR_073451.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007526.mm.1 |
NR_073452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007526.mm.1 |
TC0X00001178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007528.mm.1 |
NM_001271809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007528.mm.1 |
NM_001271810.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007528.mm.1 |
NR_073451.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007528.mm.1 |
NR_073452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007528.mm.1 |
TC0X00001178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007545.mm.1 |
NM_001271809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007545.mm.1 |
NR_073451.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007545.mm.1 |
TC0X00001178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007546.mm.1 |
NM_001271809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007546.mm.1 |
NM_001271810.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007546.mm.1 |
NR_073451.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007546.mm.1 |
NR_073452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007546.mm.1 |
TC0X00001178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007551.mm.1 |
NM_001271809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007551.mm.1 |
NM_001271810.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007551.mm.1 |
NR_073451.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007551.mm.1 |
NR_073452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007551.mm.1 |
TC0X00001178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00007541.mm.1 |
NM_001271809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007541.mm.1 |
NM_001271810.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007541.mm.1 |
NR_073451.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007541.mm.1 |
NR_073452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00007541.mm.1 |
TC0X00001178.mm.1 |
MTA TC |