Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104055.1 | 15 - 16 | 2464 | 89 | ||
GenBank mRNA | BC056456.1 | - | 15 - 16 | 2463 | 89 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ring finger protein 214 | Rnf214 | D130054N24Rik | -20.1kb | Anti-sense | Chr. 9 - 45863532 - 45906906 on NCBI Build GRCm38 |
Downstream | transgelin | Tagln |
Sm22, Sm22a, Ws310 |
2.0kb | Anti-sense | Chr. 9 - 45929628 - 45936058 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |