Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011337.2 | NP_035467.1 | 1 - 2 | 179 | 81 | |
GenBank mRNA | AA895994.1 | - | 1 - 2 | 178 | 81 | |
AF065939.1 | - | 1 - 2 | 104 | 81 | ||
AF065940.1 | - | 1 - 2 | 104 | 81 | ||
AF065941.1 | - | 1 - 2 | 104 | 81 | ||
AF065942.1 | - | 1 - 2 | 104 | 81 | ||
AF065943.1 | - | 1 - 2 | 104 | 81 | ||
AK150590.1 | - | 1 - 2 | 159 | 81 | ||
AK150634.1 | - | 1 - 2 | 159 | 81 | ||
AK150698.1 | - | 1 - 2 | 159 | 81 | ||
AK151581.1 | - | 1 - 2 | 159 | 81 | ||
AK152648.1 | - | 1 - 2 | 159 | 81 | ||
BC111443.1 | - | 1 - 2 | 103 | 40041119 | 81 | |
EU366956.1 | - | 1 - 2 | 78 | 81 | ||
J04491.1 | - | 1 - 2 | 125 | 81 | ||
X12531.1 | - | 1 - 2 | 148 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |