Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003221.4 | XP_004003270.2 | 1 - 2 | 375 | 97 | |
XM_027959067.1 | XP_027814868.1 | 1 - 2 | 405 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |