Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971422.1 | XP_027827223.1 | 1 - 2 | 753 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |