Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004397.3 | XP_004004446.2 | 2 - 3 | 454 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |