Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958945.1 | XP_027814746.1 | 2 - 3 | 407 | 68 | |
XM_027958946.1 | XP_027814747.1 | 2 - 3 | 404 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |