Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT4H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012398.4 | XP_004012447.1 | 2 - 3 | 473 | 72 | |
XM_027975360.1 | XP_027831161.1 | 1 - 2 | 151 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |