Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDF2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956872.1 | XP_027812673.1 | 2 - 3 | 426 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |