Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012174796.2 | XP_012030186.2 | 3 - 4 | 309 | 70 | |
XM_027967616.1 | XP_027823417.1 | 3 - 4 | 357 | 70 |
Gene Symbol: BLOC1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012174798.3 | XP_012030188.1 | 3 - 4 | 397 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |