Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142887.1 | NP_001136359.1 | 1 - 2 | 149 | 83 | |
GenBank mRNA | FJ444829.1 | - | 1 - 2 | 149 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |