Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020230.4 | XP_004020279.3 | 2 - 3 | 216 | 148 | |
XM_027960273.1 | XP_027816074.1 | 2 - 3 | 240 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |