Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966316.1 | XP_027822117.1 | 2 - 3 | 331 | 68 | |
XM_027966317.1 | XP_027822118.1 | 2 - 3 | 334 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |