Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018292.4 | XP_004018341.2 | 2 - 3 | 644 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |