Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018503.4 | XP_004018552.3 | 1 - 2 | 427 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |