Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973654.1 | XP_027829455.1 | 9 - 10 | 1176 | 70 | |
XM_027973656.1 | XP_027829457.1 | 9 - 10 | 1071 | 70 |