Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003320.4 | XP_004003369.1 | 12 - 13 | 1847 | 62 | |
XM_015092650.2 | XP_014948136.1 | 15 - 16 | 4157 | 62 | ||
XM_015092651.2 | XP_014948137.1 | 15 - 16 | 4190 | 62 | ||
XM_027960179.1 | XP_027815980.1 | 12 - 13 | 1408 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |