Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009215.1 | NP_001009215.1 | 1 - 2 | 573 | 73 | |
GenBank mRNA | AF108210.1 | - | 1 - 2 | 573 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Ovis aries |