Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021970.4 | XP_004022019.1 | 1 - 2 | 5141 | 102 | |
XM_027963624.1 | XP_027819425.1 | 2 - 3 | 815 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |