Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010717.4 | XP_004010766.1 | 2 - 3 | 226 | 100 | |
XM_027971920.1 | XP_027827721.1 | 1 - 2 | 10848 | 100 | ||
XM_027971921.1 | XP_027827722.1 | 1 - 2 | 10848 | 100 | ||
XM_027971922.1 | XP_027827723.1 | 1 - 2 | 10847 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |