Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015096335.2 | XP_014951821.2 | 1 - 2 | 577 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |