Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003186.4 | XP_004003235.1 | 7 - 8 | 1698 | 65 | |
XM_012099879.3 | XP_011955269.1 | 7 - 8 | 1688 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |