Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097427.2 | XP_014952913.2 | 1 - 2 | 444 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |