Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PJVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004545.4 | XP_004004594.1 | 3 - 4 | 542 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |