Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127289.1 | NP_001120761.1 | 2 - 3 | 412 | 83 | |
XM_027958330.1 | XP_027814131.1 | 2 - 3 | 414 | 83 | ||
GenBank mRNA | EU626222.1 | - | 2 - 3 | 412 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |