Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287473.1 | NP_001274402.1 | 2 - 3 | 518 | 127 | |
XM_004009046.4 | XP_004009095.1 | 3 - 4 | 730 | 127 | ||
XM_012177580.3 | XP_012032970.1 | 3 - 4 | 726 | 127 | ||
GenBank mRNA | KC539855.1 | - | 2 - 3 | 518 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |