Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009456.1 | NP_001009456.1 | 6 - 7 | 729 | 106 | |
XM_027969199.1 | XP_027825000.1 | 6 - 7 | 632 | 106 | ||
GenBank mRNA | AY327118.1 | - | 6 - 7 | 729 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |