Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127290.1 | NP_001120762.1 | 4 - 5 | 718 | 63 | |
GenBank mRNA | EU626221.1 | - | 4 - 5 | 718 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |