Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136491.1 | NP_001129963.1 | 2 - 3 | 342 | 96 | |
GenBank mRNA | AF267158.1 | - | 1 - 2 | 79 | 96 | |
FJ211194.1 | - | 2 - 3 | 342 | 96 |
Gene Symbol: LOC114118377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979143.1 | XP_027834944.1 | 2 - 3 | 403 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |