Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104933.1 | NP_001098403.1 | 6 - 7 | 1017 | 74 | |
GenBank mRNA | EF660331.1 | - | 6 - 7 | 1017 | 74 |