Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127283.1 | NP_001120755.1 | 2 - 3 | 172 | 127 | |
XM_015096239.2 | XP_014951725.1 | 2 - 3 | 393 | 127 | ||
XM_027970714.1 | XP_027826515.1 | 2 - 3 | 393 | 127 | ||
GenBank mRNA | EU626203.1 | - | 2 - 3 | 172 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |